Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC27A3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548584
Description
SLC27A3 has acyl-CoA ligase activity for long-chain and very-long-chain fatty acids.SLC27A3 does not exhibit fatty acid transport activity.
Specifications
SLC27A3 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q5K4L6 | |
SLC27A3 | |
KLH conjugated synthetic peptide between 124-152 amino acids from the N-terminal region of human SLC27A3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SLC27A3 | |
Long-chain fatty acid transport protein 3, FATP-3, Fatty acid transport protein 3, 621-, Solute carrier family 27 member 3, Very long-chain acyl-CoA synthetase homolog 3, VLCS-3, SLC27A3, ACSVL3, FATP3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11000 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction