Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC27A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 129431AP150UL

Description
SLC27A3 has acyl-CoA ligase activity for long-chain and very-long-chain fatty acids.SLC27A3 does not exhibit fatty acid transport activity.Specifications
SLC27A3 | |
Polyclonal | |
Unconjugated | |
SLC27A3 | |
ACSVL3, FATP 3, FATP3, Fatty acid transport protein 3, SLC27A3, UNQ367/PRO703, VLCS 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11000, 26568, 295219 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88561, Q5K4L6 | |
Slc27a3 | |
SLC27A3 Fusion Protein Ag4015 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction