Learn More
Invitrogen™ SLC27A4 Monoclonal Antibody (342142)

Description
This antibody stains human FATP4-transfected cells but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | SLC27A4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 342142 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | SLC27A4 |
| Gene Accession No. | Q6P1M0 |
| Gene Alias | ACSVL4; BB144259; Fatp4; FATP-4; fatty acid transport protein 4; IPS; Long-chain fatty acid transport protein 4; SLC27A4; solute carrier family 27 (fatty acid transporter), member 4; solute carrier family 27 member 4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.