Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC27A4 Recombinant Rabbit Monoclonal Antibody (8D6S1)
GREENER_CHOICE

Catalog No. FEMA551392
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551392 Supplier Invitrogen™ Supplier No. MA551392
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: RVFIKTIRRD IFGGLVLLKV KAKVRQCLQE RRTVPILFAS TVRRHPDKTA LIFEGTDTHW TFRQLDEYSS SVANFLQARG LASGDVAAIF MENRNEFVGL WLGMAKLGVE AALINTNLRR DALLHCLTTS RARALVFGSE MASAICEVHA SLDPSLSLFC SGSWEPGAVP PSTEHLDPLL KDAPKHLPSC PDKGF.

This gene encodes a member of a family of fatty acid transport proteins, which are involved in translocation of long-chain fatty acids cross the plasma membrane. This protein is expressed at high levels on the apical side of mature enterocytes in the small intestine, and appears to be the principal fatty acid transporter in enterocytes. Clinical studies suggest this gene as a candidate gene for the insulin resistance syndrome. Mutations in this gene have been associated with ichthyosis prematurity syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC27A4
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 8D6S1
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SLC27A4
Gene Accession No. Q6P1M0, Q91VE0
Gene Alias ACSVL4; BB144259; Fatp4; FATP-4; fatty acid transport protein 4; IPS; Long-chain fatty acid transport protein 4; SLC27A4; solute carrier family 27 (fatty acid transporter), member 4; solute carrier family 27 member 4
Gene Symbols SLC27A4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 43-237 of human FATP4/SLC27A4 (NP_005085.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10999, 26569
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.