Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC27A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582292
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582292 Supplier Invitrogen™ Supplier No. PA582292
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RVLVVDPDLR ESLEEILPKL QAENIRCFYL SHTSPTPGVG ALGAALDAAP SHPVPADLRA GITWRSPALF IYTSGTTGLP KPAILTHERV LQMSKMLSLS GATADDVVYT VLPLYHVMGL VVG Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 75%.

The protein encoded by this gene is an isozyme of very long-chain acyl-CoA synthetase (VLCS). It is capable of activating very long-chain fatty-acids containing 24- and 26-carbons. It is expressed in liver and associated with endoplasmic reticulum but not with peroxisomes. Its primary role is in fatty acid elongation or complex lipid synthesis rather than in degradation. This gene has a mouse ortholog.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC27A5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC27A5
Gene Accession No. Q9Y2P5
Gene Alias ACSB; ACSVL6; BA-CoA ligase; BACS; BAL; bile acid CoA ligase; bile acid-CoA ligase; Bile acyl-CoA synthetase; cholate--CoA ligase; FACVL3; FATP5; FATP-5; Fatty acid transport protein 5; fatty-acid-Coenzyme A ligase, very long-chain 3; FLJ22987; rBAL-1; Slc27a5; solute carrier family 27 (fatty acid transporter), member 5; solute carrier family 27 member 5; ve; Very long-chain acyl-CoA synthetase homolog 2; very long-chain acyl-CoA synthetase-related protein; very long-chain acyl-Coenzyme A dehydrogenase synthase related; Vlacsr; VLACS-related; VLCSH2; VLCS-H2
Gene Symbols SLC27A5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC27A5. Recombinant protein control fragment (Product #RP-89589).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10998
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.