Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC27A6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113556
Description
SLC27A6 Polyclonal Antibody for Western Blot, ELISA
The solute carrier family 27 (fatty acid transporter), member 6 (SLC27A6) is a member of the fatty acid transport protein family (FATP), which are involved in the uptake of long-chain fatty acids. SLC27A6 is principally expressed in the heart on the sarcolemma and colocalizes with CD36, another molecule involved in the uptake of long chain fatty acids.
Specifications
SLC27A6 | |
Polyclonal | |
Unconjugated | |
SLC27A6 | |
4732438L20Rik; ACSVL2; acyl-CoA synthetase very long chain family, member 2; FACVL2; FATP1; FATP6; FATP-6; Fatty-acid-coenzyme A ligase, very long-chain 2; long-chain fatty acid transport protein 6; SLC27A6; solute carrier family 27 (fatty acid transporter), member 6; solute carrier family 27 member 6; Very long-chain acyl-CoA synthetase homolog 1; VLCSH1; VLCS-H1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28965 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
0.57 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y2P4 | |
SLC27A6 | |
Recombinant Human Long-chain fatty acid transport protein 6 protein (370-619AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction