Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC28A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139830
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139830 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139830 Supplier Invitrogen™ Supplier No. PA5139830
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: MENDPSRRRE SISLTPVAKG LENMGADFLE SLEEGQLPRS DLSPAEIRSS WSEAAPKPFS RWRNLQPA Highest antigen sequence indentity to the following orthologs: Mouse - 65%, Rat - 59%.

Nucleosides play a role in signaling in several physiologic systems, and synthetic analogs of natural nucleosides are often used to treat neoplastic and viral diseases. Plasma membrane transport of nucleosides is mediated by equilibrative and concentrative nucleoside transporters, which may have specificity for purines or pyrimidines. The deduced human 650 amino acid concentrative nucleoside transporter 1 (CNT1) protein is 83% identical to the rat protein and is expressed in the intestine, kidney and liver. CNT1, also designated solute carrier family 28 (sodium-coupled nucleoside transporter), member 1 (SLC28A1), expedites sodium-dependent fluxes of uridine, azidodeoxythymidine (AZT) and adenosine, but not of guanosine or deoxyadenosine, which undergo net renal secretion. CNT1 activity may serve as a putative mechanism for renal reabsorption of physiologic nucleosides and synthetic nucleoside drugs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC28A1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc28a1
Gene Accession No. O00337
Gene Alias CNT 1; CNT1; Concentrative nucleoside transporter 1; HCNT1; Na(+)/nucleoside cotransporter 1; SLC28A1; sodium/nucleoside cotransporter 1; Sodium-coupled nucleoside transporter 1; solute carrier family 28 (concentrative nucleoside transporter), member 1; solute carrier family 28 (sodium-coupled nucleoside transporter), member 1; solute carrier family 28 member 1; solute carrier family 28, member 1
Gene Symbols Slc28a1
Host Species Rabbit
Immunogen Recombinant protein fragment of Human SLC28A1 (Product #RP-90997)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9154
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.