Learn More
Invitrogen™ SLC28A2 Polyclonal Antibody

Description
Immunogen sequence: MEKASGRQSI ALSTVETGTV NLGLELMEKE VEPEGSKRTD AQGHSLGDGL GPSTYQRRSR WPFSKARSFC KTHASLFKK Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 51%.
Specifications
Specifications
| Antigen | SLC28A2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SLC28A2 |
| Gene Accession No. | O43868 |
| Gene Alias | 2010208B10Rik; B430217P18; BB152493; CNT 2; CNT2; CNT-2; Concentrative nucleoside transporter 2; HCNT2; HsT17153; Na(+)/nucleoside cotransporter 2; purine-selective Na+ nucleoside cotransporter; rCNT2; Slc28a2; sodium/nucleoside cotransporter 2; sodium/purine nucleoside cotransporter; Sodium/purine nucleoside co-transporter; Sodium-coupled nucleoside transporter 2; solute carrier family 28 (concentrative nucleoside transporter), member 2; solute carrier family 28 (sodium-coupled nucleoside transporter), member 2; solute carrier family 28 member 2; solute carrier family 28, member 2; SPNT; SPNT1 |
| Gene Symbols | SLC28A2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.