Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC28A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563151
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563151 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563151 Supplier Invitrogen™ Supplier No. PA563151
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GAEADCVSFP NTSFTNRTYE TYMCCRGLFQ STSLNGTNPP SFSGPWEDKE FSAMALTNCC GFYNNTVCA Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 74%.

The concentrative nucleoside transporter (CNT) family comprises three members: CNT1, CNT2 and CNT3. CNT2 participates in the absorption and disposition of endogenous nucleosides and mediates the first step of nucleotide biosynthesis. CNT2 levels are highly dependent on insulin (but not glucose) concentration, and the protein is under the control of the Adenosine 1 receptor. CNT family members are imperative in the response of cells to a variety of anticancer and antiviral nucleoside analogs, as the CNT proteins modulate their entry into target tissues. Increasing evidence also suggests that CNT2 may have a role in energy metabolism because activation of CNT2 relies on the opening of ATP-sensitive K+ channels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC28A2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC28A2
Gene Accession No. O43868
Gene Alias 2010208B10Rik; B430217P18; BB152493; CNT 2; CNT2; CNT-2; Concentrative nucleoside transporter 2; HCNT2; HsT17153; Na(+)/nucleoside cotransporter 2; purine-selective Na+ nucleoside cotransporter; rCNT2; Slc28a2; sodium/nucleoside cotransporter 2; sodium/purine nucleoside cotransporter; Sodium/purine nucleoside co-transporter; Sodium-coupled nucleoside transporter 2; solute carrier family 28 (concentrative nucleoside transporter), member 2; solute carrier family 28 (sodium-coupled nucleoside transporter), member 2; solute carrier family 28 member 2; solute carrier family 28, member 2; SPNT; SPNT1
Gene Symbols SLC28A2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC28A2. Recombinant protein control fragment (Product #RP-103224).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9153
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.