Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC28A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539468
Description
The concentrative nucleoside transporter (CNT) family comprises three members: CNT1, CNT2 and CNT3. CNT2 participates in the absorption and disposition of endogenous nucleosides and mediates the first step of nucleotide biosynthesis. CNT2 levels are highly dependent on insulin (but not glucose) concentration, and the protein is under the control of the Adenosine 1 receptor. CNT family members are imperative in the response of cells to a variety of anticancer and antiviral nucleoside analogs, as the CNT proteins modulate their entry into target tissues. Increasing evidence also suggests that CNT2 may have a role in energy metabolism because activation of CNT2 relies on the opening of ATP-sensitive K+ channels.
Specifications
SLC28A2 | |
Polyclonal | |
Unconjugated | |
SLC28A2 | |
Sodium/nucleoside cotransporter 2, Concentrative nucleoside transporter 2, CNT 2, hCNT2, Na(+)/nucleoside cotransporter 2, Sodium-coupled nucleoside transporter 2, Sodium/purine nucleoside co-transporter, SPNT, Solute carrier family 28 member 2, CNT2, CNT-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
60423, 9153 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O43868, Q62773 | |
SLC28A2 | |
A synthetic peptide derived from the internal region of human SLC28A2 | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction