Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC29A3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281829
Description
SLC29A3 Polyclonal specifically detects SLC29A3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SLC29A3 | |
Polyclonal | |
Unconjugated | |
PBS | |
SLC29A3 | |
SLC29A3 antibody was raised against a 19 amino acid peptide near the amino terminus of human SLC29A3. The immunogen is located within the first 50 amino acids of SLC29A3. Amino Acid Squence: LEKLLDRPPPGLQRPEDRF | |
Affinity Purified | |
RUO | |
55315 | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin | |
equilibrative nucleoside transporter 3, ENT3, HCLAP, HJCD, PHID, solute carrier family 29 (nucleoside transporters), member 3 | |
Rabbit | |
51 kDa | |
0.1 mg | |
Primary | |
SLC29A3 antibody is human, mouse and rat reactive. At least two isoforms of SLC29A3 are known to exist; this antibody will detect both isoforms. SLC29A3 antibody is predicted to not cross-react with other SLC29 proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction