Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC29A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563045
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563045 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563045 Supplier Invitrogen™ Supplier No. PA563045
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YARYYMRPVL AAHVFSGEEE LPQDSLSAPS VASRFIDSHT PPLRPILKKT Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 62%.

SLC29A3 is a member of the equilibrative nucleoside transporter family which plays a key role in nucleoside and nucleobase uptake for salvage pathways of nucleotide synthesis. SLC29A3 is a transmembrane glycoprotein that localizes to the lysosomal membrane and is a broad selectivity, low affinity nucleoside transporter. Mutations in the SLC29A3 gene have been associated with H syndrome, which is characterized by cutaneous hyperpigmentation and hypertrichosis, hepatosplenomegaly, heart anomalies, and hypogonadism. A related disorder, PHID (pigmented hypertrichosis with insulin-dependent diabetes mellitus), has also been associated with mutations at this locus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC29A3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC29A3
Gene Accession No. Q9BZD2
Gene Alias 4933435C21Rik; AW987637; Ent3; Equilibrative nucleoside transporter 3; HCLAP; hENT3; HJCD; mENT3; PHID; Slc29a3; solute carrier family 29 (equilibrative nucleoside transporter), member 3; solute carrier family 29 (nucleoside transporters), member 3; solute carrier family 29 member 3; UNQ717/PRO1380
Gene Symbols SLC29A3
Host Species Rabbit
Immunogen Recombinant protein fragment of Human SLC29A3 (Product #RP-95808)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55315
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.