Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC29A4 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241314PCP
Description
SLC29A4 Polyclonal antibody specifically detects SLC29A4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| SLC29A4 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| ENT4PMAT, equilibrative nucleoside transporter 4, FLJ34923, hENT4, Plasma membrane monoamine transporter, solute carrier family 29 (nucleoside transporters), member 4, Solute carrier family 29 member 4 | |
| Antibody was raised against an 18 amino acid peptide near the center of human SLC29A4. The immunogen is located within amino acids 310 - 360 of SLC29A4. Amino Acid Squence: MRFDVPRPRVQRSWPTFR | |
| 0.1 mL | |
| Phospho Specific | |
| 222962 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction