Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC2A1 Rabbit anti-Human, Unconjugated, Clone: GLUT1/3132R, Abnova™

Catalog No. 89339604 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-604 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-604 Supplier Abnova Corporation Supplier No. RAB00625
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

SLC2A1 Monoclonal antibody specifically detects SLC2A1 in Human samples. It is validated for ELISA, IHC-P, Flow Cyt.

Specifications

Antigen SLC2A1
Applications ELISA, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Monoclonal
Clone GLUT1/3132R
Conjugate Unconjugated
Description solute carrier family 2 (facilitated glucose transporter), member 1
Gene SLC2A1
Gene Alias DYT17, DYT18, GLUT, GLUT1, MGC141895, MGC141896, PED
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 203-305 of human SLC2A1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6513
Target Species Human
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.