Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC2A11 Polyclonal Antibody
SLC2A11 Polyclonal Antibody
Supplier: Thermo Scientific PA550503
Description
SLC2A11 Polyclonal Antibody for IHC (P)

Specifications
SLC2A11 | |
Polyclonal | |
Unconjugated | |
SLC2A11 | |
facilitative glucose transporter GLUT11; glucose transporter protein 10; glucose transporter protein 11; glucose transporter type 10; glucose transporter type 11; glucose transporter-like protein XI; GLUT10; GLUT11; GLUT-11; gtr11; SLC2A11; solute carrier family 2 (facilitated glucose transporter), member 11; solute carrier family 2 member 11; solute carrier family 2, facilitated glucose transporter member 11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
66035 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9BYW1 | |
SLC2A11 | |
Synthetic peptide corresponding to residues near the C terminal of human solute carrier family 2 (facilitated glucose transporter), member 11. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction