Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC2A11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560174
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560174 Supplier Invitrogen™ Supplier No. PA560174
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTLHIQEFTN ETWQARTGEP LPDHL Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 54%.

Solute carrier family 2, facilitated glucose transporter member 11 (SLC2A11) also known as glucose transporter type 10/11 (GLUT-10/11) is a protein that in humans is encoded by the SLC2A11 gene. SLC2A11 belongs to a family of plasma membrane proteins that mediate transport of sugars across the membrane by facilitative diffusion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC2A11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC2A11
Gene Accession No. Q9BYW1
Gene Alias facilitative glucose transporter GLUT11; glucose transporter protein 10; glucose transporter protein 11; glucose transporter type 10; glucose transporter type 11; glucose transporter-like protein XI; GLUT10; GLUT11; GLUT-11; gtr11; SLC2A11; solute carrier family 2 (facilitated glucose transporter), member 11; solute carrier family 2 member 11; solute carrier family 2, facilitated glucose transporter member 11
Gene Symbols SLC2A11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC2A11. Recombinant protein control fragment (Product #RP-91550).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66035
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.