Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC2A11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567153
Description
Immunogen sequence: KTFQEISKELH RLNFPRRAQG PTWRSLEVIQ STEL Highest antigen sequence identity to the following orthologs - mouse 43%, rat 40%.
Solute carrier family 2, facilitated glucose transporter member 11 (SLC2A11) also known as glucose transporter type 10/11 (GLUT-10/11) is a protein that in humans is encoded by the SLC2A11 gene. SLC2A11 belongs to a family of plasma membrane proteins that mediate transport of sugars across the membrane by facilitative diffusion.
Specifications
SLC2A11 | |
Polyclonal | |
Unconjugated | |
SLC2A11 | |
facilitative glucose transporter GLUT11; glucose transporter protein 10; glucose transporter protein 11; glucose transporter type 10; glucose transporter type 11; glucose transporter-like protein XI; GLUT10; GLUT11; GLUT-11; gtr11; SLC2A11; solute carrier family 2 (facilitated glucose transporter), member 11; solute carrier family 2 member 11; solute carrier family 2, facilitated glucose transporter member 11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
66035 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BYW1 | |
SLC2A11 | |
Recombinant Human SLC2A11. Recombinant protein control fragment (Product #RP-107798). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction