Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC2A12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556789
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556789 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556789 Supplier Invitrogen™ Supplier No. PA556789
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSPRFLVMKG QEGAASKVLG RLRALSDTTE ELTVIKSSLK DEYQYSFWDL FRSKDNMRTR Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

Solute carrier family 2, facilitated glucose transporter member 12 is a protein that in humans is encoded by the SLC2A12 gene. SLC2A12 belongs to a family of transporters that catalyze the uptake of sugars through facilitated diffusion. This family of transporters show conservation of 12 transmembrane helices as well as functionally significant amino acid residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC2A12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC2A12
Gene Accession No. Q8TD20
Gene Alias facilitated glucose transporter; glucose transporter type 12; GLUT12; GLUT-12; GLUT8; SLC2A12; solute carrier family 2 (facilitated glucose transporter), member 12; solute carrier family 2 member 12; solute carrier family 2, facilitated glucose transporter member 12; solute carrier family 2, member 12
Gene Symbols SLC2A12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC2A12. Recombinant protein control fragment (Product #RP-94897).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154091
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.