Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC2A12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568915
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568915 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568915 Supplier Invitrogen™ Supplier No. PA568915
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 93%; Goat: 86%; Horse: 86%; Human: 86%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 93%.

Solute carrier family 2, facilitated glucose transporter member 12 is a protein that in humans is encoded by the SLC2A12 gene. SLC2A12 belongs to a family of transporters that catalyze the uptake of sugars through facilitated diffusion. This family of transporters show conservation of 12 transmembrane helices as well as functionally significant amino acid residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC2A12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC2A12
Gene Accession No. Q8BFW9
Gene Alias facilitated glucose transporter; glucose transporter type 12; GLUT12; GLUT-12; GLUT8; SLC2A12; solute carrier family 2 (facilitated glucose transporter), member 12; solute carrier family 2 member 12; solute carrier family 2, facilitated glucose transporter member 12; solute carrier family 2, member 12
Gene Symbols SLC2A12
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse SLC2A12 (aa 551-600).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353169
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.