Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC2A4RG Monoclonal Antibody (OTI2A5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA810377

Description
Transcription factor involved in SLC2A4 and HD gene transactivation. Binds to the consensus sequence 5′-GCCGGCG-3′.Specifications
SLC2A4RG | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9NR83 | |
SLC2A4RG | |
Human recombit protein fragment corresponding to amino acids 1-259 of human SLC2A4RG produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI2A5 | |
Unconjugated | |
SLC2A4RG | |
GEF, HDBP-1, HDBP1, Si-1-2, Si-1-2-19 | |
Mouse | |
Affinity Chromatography | |
RUO | |
56731 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction