Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC2A4RG Monoclonal Antibody (OTI4B6), OriGene
SDP

Catalog No. 502544024
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4024 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4024 Supplier OriGene Supplier No. CF810292
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC2A4RG Monoclonal antibody specifically detects SLC2A4RG in Human samples. It is validated for Western Blot

Specifications

Antigen SLC2A4RG
Applications Western Blot
Classification Monoclonal
Clone OTI4B6
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene TRAF5
Gene Accession No. O00463
Gene Alias GEF; GLUT4 enhancer factor; HDBP1; HDBP-1; Huntington disease gene regulatory region-binding protein 1; Huntington's disease gene regulatory region-binding protein 1; Si-1-2; Si-1-2-19; SLC2A4 regulator; SLC2A4RG
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 334-557 of human TRAF5 produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7188
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.