Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC2A4RG Monoclonal Antibody (OTI8A3), TrueMAB™, OriGene
SDP

Catalog No. 501683362
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3362 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3362 Supplier OriGene Supplier No. TA810378
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transcription factor involved in SLC2A4 and HD gene transactivation. Binds to the consensus sequence 5′-GCCGGCG-3′.

Specifications

Antigen SLC2A4RG
Applications Western Blot
Classification Monoclonal
Clone OTI8A3
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SLC2A4RG
Gene Accession No. Q9NR83
Gene Alias GEF, HDBP-1, HDBP1, Si-1-2, Si-1-2-19
Gene Symbols SLC2A4RG
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-259 of human SLC2A4RG produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56731
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.