Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC2A4RG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595892
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595892 Supplier Invitrogen™ Supplier No. PA595892
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARLDEVMAAA ALTSLSTSPL LLGAPVAAFS PEPGLEPWKE ALVRPPGSYS SSSNSGDWGW DLASDQSSPS TPSPPLPPEA AHFLFGEPTL RKRKSPAQVM F.

Transcription factor involved in SLC2A4 and HD gene transactivation. Binds to the consensus sequence 5'-GCCGGCG-3'.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC2A4RG
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Alias GEF; GLUT4 enhancer factor; HDBP1; HDBP-1; Huntington disease gene regulatory region-binding protein 1; Huntington's disease gene regulatory region-binding protein 1; Si-1-2; Si-1-2-19; SLC2A4 regulator; SLC2A4RG
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human SLC2A4RG (NP_0644462).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.