Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC2A4RG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566553
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566553 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566553 Supplier Invitrogen™ Supplier No. PA566553
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVLSTVANPQS CHSDRVYQGC LTPARLEPQP TEVGACPPAL SSRIGVTLRK P Highest antigen sequence identity to the following orthologs - mouse 29%, rat 33%.

Transcription factor involved in SLC2A4 and HD gene transactivation. Binds to the consensus sequence 5'-GCCGGCG-3'.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC2A4RG
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC2A4RG
Gene Accession No. Q9NR83
Gene Alias GEF; GLUT4 enhancer factor; HDBP1; HDBP-1; Huntington disease gene regulatory region-binding protein 1; Huntington's disease gene regulatory region-binding protein 1; Si-1-2; Si-1-2-19; SLC2A4 regulator; SLC2A4RG
Gene Symbols SLC2A4RG
Host Species Rabbit
Immunogen Recombinant Human SLC2A4RG. Recombinant protein control fragment (Product #RP-107213).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56731
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.