Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC2A4RG Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989083
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9083 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9083 Supplier Bioss Supplier No. BS9457R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transcription factor involved in SLC2A4 and HD gene transactivation. Binds to the consensus sequence 5′-GCCGGCG-3′.

Specifications

Antigen SLC2A4RG
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLC2A4RG
Gene Accession No. Q9NR83
Gene Alias GEF; GLUT4 enhancer factor; HDBP1; HDBP-1; Huntington disease gene regulatory region-binding protein 1; Huntington′s disease gene regulatory region-binding protein 1; Si-1-2; Si-1-2-19; SLC2A4 regulator; SLC2A4RG
Gene Symbols SLC2A4RG
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SLC2A4RG/GLUT4 enhancer factor.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56731
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.