Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC2A5 Monoclonal Antibody (14C8)
Mouse Monoclonal Antibody
$235.50 - $628.50
Specifications
Antigen | SLC2A5 |
---|---|
Clone | 14C8 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Description
This antibody requires overnight incubation in a Western blot for optimal performance. Do not use this antibody in a Fast Western blotting procedure.
Cytochalasin B-sensitive carrier. Seems to function primarily as a fructose transporter.Specifications
SLC2A5 | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 1 mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
P22732, Q9WV38 | |
56485, 6518 | |
Protein expressed in 293T cell transfected with human SLC2A5 expression vector. | |
Antibody |
14C8 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG1 | |
Human, Mouse | |
SLC2A5 | |
AI526984; Fructose transporter; glucose transporter type 5, small intestine; glucose transporter-like protein 5; GLUT5; GLUT-5; RP5-963K15.1; SLC2A5; Slc5a; solute carrier family 2 (facilitated glucose transporter), member 5; solute carrier family 2 (facilitated glucose/fructose transporter), member 5; solute carrier family 2 member 5; solute carrier family 2, facilitated glucose transporter member 5; solute carrier family 2, member 5; testicular tissue protein Li 81 | |
SLC2A5 | |
Primary | |
Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title