Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC2A5 Mouse anti-Human, Clone: OTI9G8, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA500604

Description
SLC2A5 (GLUT5) is a cytochalasin B-sensitive carrier. It seems to function primarily as a fructose transporter.Specifications
SLC2A5 | |
Monoclonal | |
0.56 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P22732 | |
SLC2A5 | |
Full length human recombit protein of human SLC2A5 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI9G8 | |
Unconjugated | |
SLC2A5 | |
GLUT-5, GLUT5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6518 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction