Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC2A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542100
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542100 Supplier Invitrogen™ Supplier No. PA542100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ADQIYLSAGV PEEHVQYVTA GTGAVNVVMT FCAVFVVELL GRRLLLLLGF Sequence homology: Cow: 85%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Sheep: 85%.

Cytochalasin B-sensitive carrier. Seems to function primarily as a fructose transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC2A5
Applications Western Blot, Immunocytochemistry, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC2A5
Gene Accession No. P22732, Q9WV38
Gene Alias AI526984; Fructose transporter; glucose transporter type 5, small intestine; glucose transporter-like protein 5; GLUT5; GLUT-5; RP5-963K15.1; SLC2A5; Slc5a; solute carrier family 2 (facilitated glucose transporter), member 5; solute carrier family 2 (facilitated glucose/fructose transporter), member 5; solute carrier family 2 member 5; solute carrier family 2, facilitated glucose transporter member 5; solute carrier family 2, member 5; testicular tissue protein Li 81
Gene Symbols SLC2A5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SLC2A5.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56485, 6518
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.