Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC2A6 Monoclonal Antibody (OTI7E3)
GREENER_CHOICE

Catalog No. PIMA524979
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524979 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524979 Supplier Invitrogen™ Supplier No. MA524979
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC2A6 Monoclonal Antibody for ICC/IF, IHC (P)

Hexose transport into mammalian cells is catalyzed by a family of membrane proteins, including SLC2A6, that contain 12 transmembrane domains and a number of critical conserved residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC2A6
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI7E3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SLC2A6
Gene Accession No. Q9UGQ3
Gene Alias A330096C23; F630103L12Rik; Glucose transporter type 6; glucose transporter type 9; Glut6; GLUT-6; Glut9; GLUT-9; HSA011372; SLC2A6; solute carrier family 2 (facilitated glucose transporter), member 6; solute carrier family 2 member 6; solute carrier family 2, facilitated glucose transporter member 6
Gene Symbols SLC2A6
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human SLC2A6 expression vector.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11182
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.