Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC2A8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552877
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552877 Supplier Invitrogen™ Supplier No. PA552877
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRFLLTQHRR QEAMAALRFL WGSEQGWEDP PIGAEQSFHL ALLRQPGIYK P Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 77%.

SLC2A8 is the insulin-regulated facilitative glucose transporter.SLC2A8 binds cytochalasin B in a glucose-inhibitable manner.SLC2A8 seems to be a dual-specific sugar transporter as it is inhibitable by fructose.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC2A8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC2A8
Gene Accession No. Q9NY64
Gene Alias D2Ertd44e; Glucose transporter type 8; glucose transporter type X1; glucose transporter X1; Glut8; GLUT-8; GLUTX1; intestinal GLUT8 transporter; SLC2A8; solute carrier family 2 (facilitated glucose transporter), member 8; solute carrier family 2 member 8; solute carrier family 2, (facilitated glucose transporter) member 8; solute carrier family 2, (facilitated glucose transporter), member 8; solute carrier family 2, facilitated glucose transporter member 8
Gene Symbols SLC2A8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC2A8. Recombinant protein control fragment (Product #RP-89665).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29988
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.