Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC30A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588608
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588608 Supplier Invitrogen™ Supplier No. PA588608
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLYTTYPLLK ESALILLQTV PKQIDIRNLI KELRNVEGVE EVHELHVWQL AGSRIIATAH IKCEDPTSYM EVAKTIKDVF HNHGIHATTI QPEFASVGSK SSVVPCELAC RTQCALKQCC GTLPQAPSGK DAEKTPAVSI SCLELSNNLE KKPRRTKAEN IPAVVIEIKN MPNKQPESSL; Positive Samples: Mouse liver, Rat liver; Cellular Location: Cell membrane, Multi-pass membrane protein.

Zinc, an essential element required for cell proliferation and differentiation, plays a role in a diverse array of cellular functions, including acting as a cofactor for numerous enzymes and transcription factors and as a neuroregulator. The zinc transporter (ZnT) family regulates the supply of zinc within cells, and its members are characterized by containing six membrane-spanning domains, a large histidine-rich intracellular loop, and a C-terminal tail. ZnT-1, a ubiquitous protein of approximately 60 kDa, localizes to the plasma membrane to aid in the export of zinc out of cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC30A1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Slc30a1
Gene Accession No. Q60738, Q62720
Gene Alias AI839647; C130040I11Rik; Slc30a1; solute carrier family 30 (zinc transporter), member 1; solute carrier family 30 member 1; solute carrier family 30, member 1; zinc transporter 1; ZNT1; ZnT-1; ZnT-1 zinc transporter; ZRC1
Gene Symbols Slc30a1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 328-507 of human SLC30A1 (NP_0670172).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22782, 58976
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.