Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC30A10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549188
Description
SLC30A10 Polyclonal Antibody for Western Blot
May be involved in zinc transport out of the cell, being a zinc-efflux transporter.
Specifications
SLC30A10 | |
Polyclonal | |
Unconjugated | |
SLC30A10 | |
E130106K10Rik; Gm212; HMDPC; HMNDYT1; Manganese transporter SLC30A10; SLC30A10; Solute carrier family 30 member 10; solute carrier family 30, member 10; zinc transporter 10; zinc transporter 8; Znt10; znT-10; ZNT8; ZRC1 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
55532 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q6XR72 | |
SLC30A10 | |
KLH conjugated synthetic peptide between 331-359 amino acids of human SLC30A10. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction