Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC30A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575948
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The protein encoded by the SLC30A2 gene is a zinc transporter that acts as a homodimer. The encoded protein plays a role in secreting zinc into breast milk. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
SLC30A2 | |
Polyclonal | |
Unconjugated | |
SLC30A2 | |
6030430C21; PP12488; SLC30A2; solute carrier family 30 (zinc transporter), member 2; solute carrier family 30 member 2; solute carrier family 30, member 2; TNZD; Zinc transporter 2; ZNT2; ZnT-2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230810, 25362, 7780 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2HJ10, Q62941, Q9BRI3 | |
SLC30A2 | |
Synthetic peptide corresponding to amino acids 125-170 of Human SLC30A2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction