Learn More
SLC30A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111914
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)GSHPNSE KQRARRK Homology: Mouse - 12/14 amino acid residues identical.
The protein encoded by the SLC30A2 gene is a zinc transporter that acts as a homodimer. The encoded protein plays a role in secreting zinc into breast milk. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
SLC30A2 | |
Polyclonal | |
Unconjugated | |
SLC30A2 | |
6030430C21; PP12488; SLC30A2; Solute carrier family 30 member 2; TNZD; Zinc transporter 2; Znt2; ZnT-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
230810, 25362 | |
-20°C | |
Lyophilized |
Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q2HJ10, Q62941 | |
SLC30A2 | |
Peptide corresponding to amino acid residues 43-56 of rat ZnT2 (Intracellular, N-terminus) | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.