Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC30A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569263
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569263 Supplier Invitrogen™ Supplier No. PA569263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%.

Slc30a3 encodes a protein that is involved in accumulation of zinc in synaptic vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC30A3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Slc30a3
Gene Accession No. Q99726
Gene Alias SLC30A3; solute carrier family 30 (zinc transporter), member 3; solute carrier family 30 member 3; solute carrier family 30 protein; zinc transporter 3; zinc transporter ZnT-3; ZNT3; znT-3
Gene Symbols Slc30a3
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SLC30A3.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7781
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.