Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC30A4 Polyclonal Antibody

Catalog No. PIPA553749
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553749 Supplier Invitrogen™ Supplier No. PA553749

Rabbit Polyclonal Antibody

Immunogen sequence: PSHLNVDYIK EALMKIEDVY SVEDLNIWSL TSGKSTAIVH IQLIPGSSSK WEEVQSKANH LLLNTFGMYR CTIQLQSYRQ EVDRTCANCQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

Zinc is the second most abundant trace metal in the human body. It is an essential element, serving both a structural role, as in the formation of zinc fingers in DNA-binding proteins, and a catalytic role in metalloenzymes, such as pancreatic carboxypeptidases (e.g., MIM 114852), alkaline phosphatases (e.g., MIM 171760), various dehydrogenases, and superoxide dismutases (e.g., MIM 147450). SLC30A4, or ZNT4, belongs to the ZNT family of zinc transporters. ZNTs are involved in transporting zinc out of the cytoplasm and have similar structures, consisting of 6 transmembrane domains and a histidine-rich cytoplasmic loop (Huang and Gitschier, 1997).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC30A4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc30a4
Gene Accession No. O14863
Gene Alias dri 27 protein; Dri 27/ZnT4 protein; lethal milk protein; Lm; Slc30a4; solute carrier family 30 (zinc transporter), member 4; solute carrier family 30 (zinc transporter), member 4; zinc transporter 4; solute carrier family 30 member 4; Zinc transporter 4; ZNT4; znT-4
Gene Symbols Slc30a4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC30A4. Recombinant protein control fragment (Product #RP-91249).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7782
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.