Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC30A4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106776
Description
Antibody detects endogenous levels of total SLC30A4.
Zinc is the second most abundant trace metal in the human body. It is an essential element, serving both a structural role, as in the formation of zinc fingers in DNA-binding proteins, and a catalytic role in metalloenzymes, such as pancreatic carboxypeptidases (e.g., MIM 114852), alkaline phosphatases (e.g., MIM 171760), various dehydrogenases, and superoxide dismutases (e.g., MIM 147450). SLC30A4, or ZNT4, belongs to the ZNT family of zinc transporters. ZNTs are involved in transporting zinc out of the cytoplasm and have similar structures, consisting of 6 transmembrane domains and a histidine-rich cytoplasmic loop (Huang and Gitschier, 1997).
Specifications
SLC30A4 | |
Polyclonal | |
Unconjugated | |
Slc30a4 | |
dri 27 protein; Dri 27/ZnT4 protein; lethal milk protein; Lm; Slc30a4; solute carrier family 30 (zinc transporter), member 4; solute carrier family 30 (zinc transporter), member 4; zinc transporter 4; solute carrier family 30 member 4; Zinc transporter 4; ZNT4; znT-4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22785, 64469, 7782 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O14863, O35149, O55174 | |
Slc30a4 | |
A synthesized peptide derived from human SLC30A4(Accession O14863), corresponding to amino acid residues V228-A278. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction