Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC30A4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA580028
Description
The synthetic peptide sequence is 407-427aa, TIQLQSYRQEVDRTCANCQSS Add 0.2 mL of distilled water, will yield a concentration of 500 μg/mL.
Zinc is the second most abundant trace metal in the human body. It is an essential element, serving both a structural role, as in the formation of zinc fingers in DNA-binding proteins, and a catalytic role in metalloenzymes, such as pancreatic carboxypeptidases (e.g., MIM 114852), alkaline phosphatases (e.g., MIM 171760), various dehydrogenases, and superoxide dismutases (e.g., MIM 147450). SLC30A4, or ZNT4, belongs to the ZNT family of zinc transporters. ZNTs are involved in transporting zinc out of the cytoplasm and have similar structures, consisting of 6 transmembrane domains and a histidine-rich cytoplasmic loop (Huang and Gitschier, 1997).
Specifications
SLC30A4 | |
Polyclonal | |
PBS with 5MG BSA, 5MG BSA and 0.05MG sodium azide, 0.05MG sodium azide, 0.05MG thimerosal, 0.05MG thimerosal | |
O14863, O35149, O55174 | |
Slc30a4 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human SLC30A4. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SLC30A4 | |
Zinc transporter 4, ZnT-4, Solute carrier family 30 member 4, SLC30A4, ZNT4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22785, 64469, 7782 | |
-20°C | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction