Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC30A6 Monoclonal Antibody (2G6-G3)
GREENER_CHOICE

Catalog No. PIMA524484
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524484 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524484 Supplier Invitrogen™ Supplier No. MA524484
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MSVYSGKVLL QTTPPHVIGQ LDKLIREVST LDGVLEVRNE HFWTLGFGSL AGSVHVRIRR DANEQMVLAH VTNRLYTLVS TLTVQIFKDD WIRPALLSGP VAANVLNFSD HHVIPMPLLK GTDDLNPVTS TPAKPSSPPP EFSFNTPGKN VNPVILLNTQ TRPYGFGLNH GHTPYSSMLN QGLGVPGIGA TQGLRTGFTN IPSRYGTNNR IGQPRP.

Zinc functions as a cofactor for numerous enzymes, nuclear factors, and hormones and as an intra- and intercellular signal ion. Members of the zinc transporter (ZNT)/SLC30 subfamily of the cation diffusion facilitator family, such as SLC30A6, permit cellular efflux of zinc (Seve et al., 2004).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC30A6
Applications ELISA
Classification Monoclonal
Clone 2G6-G3
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SLC30A6
Gene Accession No. Q6NXT4
Gene Alias 9530029F08Rik; MST103; MSTP103; SLC30A6; solute carrier family 30 (zinc transporter), member 4; solute carrier family 30 (zinc transporter), member 6; solute carrier family 30 member 6; Zinc transporter 6; zinc transporter-like 3; Znt6; znT-6
Gene Symbols SLC30A6
Host Species Mouse
Immunogen SLC30A6 (AAH32525, 1 a.a. ~ 216 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55676
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.