Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC30A6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565919 Supplier Invitrogen™ Supplier No. PA565919
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGFGSLAGSVH VRIRRDANEQ MVLAHVTNRL YTLVSTLTVQ IFKDDWIRPA LLSGPVAANV LNFSDHHVIP MPLLKGTDDL NPVT Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.

Zinc functions as a cofactor for numerous enzymes, nuclear factors, and hormones and as an intra- and intercellular signal ion. Members of the zinc transporter (ZNT)/SLC30 subfamily of the cation diffusion facilitator family, such as SLC30A6, permit cellular efflux of zinc (Seve et al., 2004).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC30A6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC30A6
Gene Accession No. Q6NXT4
Gene Alias 9530029F08Rik; MST103; MSTP103; SLC30A6; solute carrier family 30 (zinc transporter), member 4; solute carrier family 30 (zinc transporter), member 6; solute carrier family 30 member 6; Zinc transporter 6; zinc transporter-like 3; Znt6; znT-6
Gene Symbols SLC30A6
Host Species Rabbit
Immunogen Recombinant Human SLC30A6. Recombinant protein control fragment (Product #RP-106597).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55676
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.