Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC30A6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571831
Description
SLC30A6 Polyclonal Antibody for Western Blot
Zinc functions as a cofactor for numerous enzymes, nuclear factors, and hormones and as an intra- and intercellular signal ion. Members of the zinc transporter (ZNT)/SLC30 subfamily of the cation diffusion facilitator family, such as SLC30A6, permit cellular efflux of zinc (Seve et al., 2004).
Specifications
SLC30A6 | |
Polyclonal | |
Unconjugated | |
SLC30A6 | |
9530029F08Rik; MST103; MSTP103; SLC30A6; solute carrier family 30 (zinc transporter), member 4; solute carrier family 30 (zinc transporter), member 6; solute carrier family 30 member 6; Zinc transporter 6; zinc transporter-like 3; Znt6; znT-6 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
210148, 55676 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q6NXT4, Q8BJM5 | |
SLC30A6 | |
KLH conjugated synthetic peptide between 282-310 amino acids from the Central region of human SLC30A6. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction