Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC30A6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588194
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588194 Supplier Invitrogen™ Supplier No. PA588194
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVLLQTTPPH VIGQLDKLIR EVSTLDGVLE VRNEHFWTLG FGSLAGSVHV RIRRDANEQM VLAHVTNRLY TLVSTLTVQI FKDDWIRPAL LSGPVAANVL NFSDHHVIPM PLLKGTDDLN PVTSTPAKPS SPPPEFSFNT PGKNVNPVIL LNTQTRPYGF GLNHGHTPYS SMLNQGLGVP GIGATQGLRT GFTNIPSRYG TNNRIGQPRP; Positive Samples: U-87MG, Mouse brain, Rat brain; Cellular Location: Golgi apparatus, Multi-pass membrane protein, trans-Golgi network membrane.

Zinc functions as a cofactor for numerous enzymes, nuclear factors, and hormones and as an intra- and intercellular signal ion. Members of the zinc transporter (ZNT)/SLC30 subfamily of the cation diffusion facilitator family, such as SLC30A6, permit cellular efflux of zinc (Seve et al., 2004).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC30A6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SLC30A6
Gene Accession No. Q6NXT4, Q8BJM5
Gene Alias 9530029F08Rik; MST103; MSTP103; SLC30A6; solute carrier family 30 (zinc transporter), member 4; solute carrier family 30 (zinc transporter), member 6; solute carrier family 30 member 6; Zinc transporter 6; zinc transporter-like 3; Znt6; znT-6
Gene Symbols SLC30A6
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 210148, 298786, 55676
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.