Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC30A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553709
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553709 Supplier Invitrogen™ Supplier No. PA553709
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HGGHGHSHGS GHGHSHSLFN GALDQAHGHV DHCHSHEVKH GAAHSHDHAH GHGHFHSHDG PSLKETTGPS RQI Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 85%.

Zinc functions as a cofactor for numerous enzymes, nuclear factors, and hormones and as an intra- and intercellular signal ion. Members of the zinc transporter (ZNT)/SLC30 subfamily of the cation diffusion facilitator family, such as SLC30A7, permit cellular efflux of zinc.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC30A7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC30A7
Gene Accession No. Q8NEW0
Gene Alias 1810059J10Rik; 2610034N15Rik; 4833428C12Rik; AI467242; Slc30a7; solute carrier family 30 (zinc transporter), member 7; solute carrier family 30 member 7; Zinc transporter 7; zinc transporter like 2; zinc transporter ZnT-7; ZNT7; znT-7; ZNTL2; Znt-like transporter 2
Gene Symbols SLC30A7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC30A7. Recombinant protein control fragment (Product #RP-91261).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148867
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.