Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Slc30A8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119426
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119426 200 μL
PIPA5121603 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119426 Supplier Invitrogen™ Supplier No. PA5119426
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Slc30A8 Polyclonal Antibody for Western Blot, IHC (P)

Slc30A8 is a zinc efflux transporter involved in the accumulation of zinc in intracellular vesicles and is expressed at a high level in the pancreas, particularly in islets of Langerhans, and co-localizes with insulin in the secretory pathway granules of the insulin-secreting INS-1 cells. Allelic variants of Slc30A8 have been identified as major genetic risk factors for the development of Type 2 diabetes, but no genetic association has been found with Type 1 diabetes. Slc30A8 expression in b-cells was found to be influenced by cytokine expression, particularly IFN-gamma and IL-1 beta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Slc30A8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SLC30A8
Gene Accession No. Q8BGG0, Q8IWU4
Gene Alias C820002P14Rik; LOW QUALITY PROTEIN: uncharacterized protein slc30a8; Slc30a8; solute carrier family 30 (zinc transporter), member 8; solute carrier family 30 member 8; wu:fe49g03; Zinc transporter 8; zinc transporter 8; LOW QUALITY PROTEIN: zinc transporter 8; zinc transporter ZnT-8; Znt8; ZnT-8
Gene Symbols SLC30A8
Host Species Rabbit
Immunogen Recombinant protein Solute Carrier Family 30 Member 8. The antigen corresponds to amino acid range 263-369 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 169026, 239436
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.