Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC30A8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584993
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584993 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584993 Supplier Invitrogen™ Supplier No. PA584993
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSAHVATAAS RDSQVVRREI AKALSKSFTM HSLTIQMESP VDQDPDCLFC EDPCD Highest antigen sequence indentity to the following orthologs: Mouse - 73%, Rat - 73%.

Slc30A8 is a zinc efflux transporter involved in the accumulation of zinc in intracellular vesicles and is expressed at a high level in the pancreas, particularly in islets of Langerhans, and co-localizes with insulin in the secretory pathway granules of the insulin-secreting INS-1 cells. Allelic variants of Slc30A8 have been identified as major genetic risk factors for the development of Type 2 diabetes, but no genetic association has been found with Type 1 diabetes. Slc30A8 expression in b-cells was found to be influenced by cytokine expression, particularly IFN-gamma and IL-1 beta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC30A8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC30A8
Gene Accession No. Q8IWU4
Gene Alias C820002P14Rik; LOW QUALITY PROTEIN: uncharacterized protein slc30a8; Slc30a8; solute carrier family 30 (zinc transporter), member 8; solute carrier family 30 member 8; wu:fe49g03; Zinc transporter 8; zinc transporter 8; LOW QUALITY PROTEIN: zinc transporter 8; zinc transporter ZnT-8; Znt8; ZnT-8
Gene Symbols SLC30A8
Host Species Rabbit
Immunogen Recombinant protein fragment of Human SLC30A8 (Product #RP-108278)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 169026
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.