Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC30A9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101996
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101996 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101996 Supplier Invitrogen™ Supplier No. PA5101996
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC30A9.

The gene corresponding to embryonic lung protein [also known as Solute carrier family 30 (Zinc transporter), member 9, SLC30A9] is likely to be an evolutionarily conserved, housekeeping gene that plays a role intimately linked with cellular replication, DNA synthesis and/or transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC30A9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC30A9
Gene Accession No. Q5IRJ6, Q6PML9
Gene Alias 2310024J23Rik; AL024256; C4orf1; expressed in human embryonic lung; GAC63; GRIP1-associated coactivator 1; GRIP1-associated coactivator 63; GRIP1-dependent nuclear receptor coactivator; HUEL; Human embryonic lung protein; Slc30a9; solute carrier family 30 (zinc transporter), member 9; solute carrier family 30 member 9; Zinc transporter 9; ZNT9; znT-9
Gene Symbols SLC30A9
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC30A9(Accession Q6PML9), corresponding to amino acid residues S171-N221.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10463, 109108, 498358
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.