Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC30A9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552043
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552043 Supplier Invitrogen™ Supplier No. PA552043
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HRCSWSSLCR LRLRCRAAAC NPSDRQEWQN LVTFGSFSNV VPCSHPYIGT LSQVKLYSTN VQKEGQGSQT LRVEKVPSFE TAEGIGAELK APLKQEPLQV RVKAVLKKRE YGSKYTQNNF ITGV Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 66%.

The gene corresponding to embryonic lung protein [also known as Solute carrier family 30 (Zinc transporter), member 9, SLC30A9] is likely to be an evolutionarily conserved, housekeeping gene that plays a role intimately linked with cellular replication, DNA synthesis and/or transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC30A9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC30A9
Gene Accession No. Q6PML9
Gene Alias 2310024J23Rik; AL024256; C4orf1; expressed in human embryonic lung; GAC63; GRIP1-associated coactivator 1; GRIP1-associated coactivator 63; GRIP1-dependent nuclear receptor coactivator; HUEL; Human embryonic lung protein; Slc30a9; solute carrier family 30 (zinc transporter), member 9; solute carrier family 30 member 9; Zinc transporter 9; ZNT9; znT-9
Gene Symbols SLC30A9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC30A9. Recombinant protein control fragment (Product #RP-102351).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10463
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.