Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC31A1/CTR1 Antibody (5E6.1B4), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP236573AF350
Description
SLC31A1/CTR1 Monoclonal specifically detects SLC31A1/CTR1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SLC31A1/CTR1 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
SLC31A1 | |
Two synthetic peptides made to internal sequences of human SLC31A1/CTR1 protein (sequences found between amino acids 50-150) [UniProtNo. O15431] | |
Primary | |
Human, Mouse | |
IgG2b κ |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
5E6.1B4 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Copper transporter 1, COPT1high affinity copper uptake protein 1, CTR1MGC75487, hCTR1copper transport 1 homolog, solute carrier family 31 (copper transporters), member 1, Solute carrier family 31 member 1 | |
Mouse | |
0.1 mL | |
1317 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction