Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC33A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584453
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584453 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584453 Supplier Invitrogen™ Supplier No. PA584453
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGGWDDSHLD SAGREGDREA LLGDTGTGDF LKAPQSFRAE LS Highest antigen sequence indentity to the following orthologs: Rat - 74%, Mouse - 71%.

SLC33A1 is required for the formation of O-acetylated (Ac) gangliosides. It is predicted to contain 6 to 10 transmembrane domains, and a leucine zipper motif in transmembrane domain III. Studies indicate that the protein is localized to the cytoplasm.The encoded protein is required for the formation of O-acetylated (Ac) gangliosides. It is predicted to contain 6 to 10 transmembrane domains, and a leucine zipper motif in transmembrane domain III. Studies indicate that the protein is localized to the cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC33A1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC33A1
Gene Accession No. O00400
Gene Alias Acatn; acetyl-CoA transporter 1; acetyl-coenzyme A transporter; acetyl-coenzyme A transporter 1; AI315656; AI788741; AT1; AT-1; CCHLND; D630022N01Rik; similar to solute carrier family 33 (acetyl-CoA transporter), member 1; SLC33A1; solute carrier family 33 (acetyl-CoA transporter), member 1; solute carrier family 33 member 1; SPG42; zgc:63693
Gene Symbols SLC33A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC33A1. Recombinant protein control fragment (Product #RP-108445).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9197
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.