Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC33A1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980451
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0451 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0451 Supplier Bioss Supplier No. BS0699R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC33A1 is required for the formation of O-acetylated (Ac) gangliosides. It is predicted to contain 6 to 10 transmembrane domains, and a leucine zipper motif in transmembrane domain III. Studies indicate that the protein is localized to the cytoplasm.The encoded protein is required for the formation of O-acetylated (Ac) gangliosides. It is predicted to contain 6 to 10 transmembrane domains, and a leucine zipper motif in transmembrane domain III. Studies indicate that the protein is localized to the cytoplasm.

Specifications

Antigen SLC33A1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLC33A1
Gene Accession No. O00400
Gene Alias Acatn; acetyl-CoA transporter 1; acetyl-coenzyme A transporter; Acetyl-coenzyme A transporter 1; AI315656; AI788741; AT1; AT-1; BOS_1131; CCHLND; D630022N01Rik; similar to solute carrier family 33 (acetyl-CoA transporter), member 1; SLC33A1; solute carrier family 33 (acetyl-CoA transporter), member 1; Solute carrier family 33 member 1; SPG42; zgc:63693
Gene Symbols SLC33A1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SLC33A1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9197
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.